VWR®, メカニカルタイマー
Mechanical timer, White, 3 min, 57×57 mm
Catalog # 609-0127
Supplier: VWR Collection
Jump to:
Product Details & Documents
This dial timer has a timing capacity of 60 minutes.
- Audible alarm
- Highly visible red arrow
Documents
609-0127
Images 360
Specifications
- Colour:White
- 説明:Mechanical timer
- Accuracy:3 min
- W×H:57×57 mm