Orders LinkContactUsLinkComponent
VWR®, Stirrer Blades with Plain Ends
VWR®, Stirrer Blades with Plain Ends
Supplier:   VWR Collection
VWR®, Stirrer Blades with Plain Ends
Supplier:  VWR Collection
Certificates
VWR®, Stirrer Blades with Plain Ends
VWR®, Stirrer Blades with Plain Ends
5 Options Available Below
Product Details & Documents
PTFE. Blades fit PTFE stirring shaft; hole Ø 6,5 mm, 3,2 mm thick. Inert and non scratch. The smooth surface prevents contamination and makes cleaning easier.
  • Excellent chemical resistance
  • Temperature resistant from −200 to 280 °C
Product Family Options
Product Information
  • Type
    高さ
    Availability
    Price
  • Stirrer blade, plain ends
    65 mm
    25 mm
    1 * 1 items
  • Stirrer blade, plain ends
    75 mm
    25 mm
    1 * 1 items
  • Stirrer blade, plain ends
    105 mm
    25 mm
    1 * 1 items
  • Stirrer blade, plain ends
    125 mm
    25 mm
    1 * 1 items
  • Stirrer blade, plain ends
    150 mm
    25 mm