VWR®, チューブローテーター
Product Details & Documents
A unique design of rotator makes this unit suitable for a wide range of mixing applications in biochemistry, molecular biology and histochemistry. The rotisserie assemblies allow for mixing tubes vertically, horizontally, or any position in between, therefore, gentle mixing, using a horizontal position is easily accommodated for applications such as hybridisation. Alternatively rotation with vertically positioned tubes produces a tumbling motion suitable for mixing samples in blood tubes. The two halves of the rotisserie can be adjusted independently of each other to increase versatility and facilitate optimisation of mixing.
- Adjustable mixing angle changed with a simple turn of the plate - no tools required
- Can be used at 4 to 65 °C in cold room or incubator
- Rotisserie assemblies available for a range of tube sizes
- Moulded housing and rotisserie plates are easy to clean
- Lightweight unit with a small footprint for easy manoeuvring and transport
: The tube rotator is supplied with a rotisserie assembly for 36×1,5/2,0 ml microtubes; other rotisseries must be ordered separately.
Documents
444-0500
Images 360
VWR® for Stirring, Mixing, Shaking And Blending
Product Literature
Specifications
- 説明:Avantor rotator, EU-plug
- Seq:01
- Shaking motion:Rotating
- Cat. No.:444-0500
- Speed:20 min⁻¹ (fixed)
- Angle:Plates freely adjustable from horizontal to vertical
- Capacity:36×1,5/2,0 ml tubes or 10×10/15 ml and 16×5/7 ml tubes or 6×50 ml tubes
- Model:Tube rotator
- W×D×H:239×127×153 mm
- Max. load:0,8 kg
- Weight:2 kg
- Operating temperature:4...65 °C