Orders LinkContactUsLinkComponent
VWR®, マイクロプレートシェーカー、高速
VWR®, マイクロプレートシェーカー、高速
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, マイクロプレートシェーカー、高速
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, マイクロプレートシェーカー、高速
2 Options Available Below
Product Details & Documents
Ideal for molecular biology applications, mechanical and chemical cell lysis, mixing tissue samples, mixing cytogenetic suspensions, and vortexing cell suspensions. Shaker is also ideal for emulsification of bi-polar viscous fluids and for mixing fluids in small diameter tubes. Specifically designed to shake and/or vortex microplates in timed or continuous modes. Shaker features programmable timed mode from 1 s to 160 hours. Speed and time are easily adjusted with the up/down arrows on the front panel.
  • Holds up to 6 microplates without stacking, up to a maximum of 48 microplates with stacking (6 in layers horizontally, up to 8 in a vertical stack). Can be used with vessels up to 127 mm high
  • Programmable digital speed control and timer
  • Programmable pulsing feature to enhance mixing action
  • Can be used at 4 to 40 °C, in an incubator or cold room
  • RS232 interface
The 279×311 mm tray assembly includes a foam pad and accepts up to 48 96-well microplates. Tray assembly height is adjustable from 3,2 mm up to 127 mm, accommodating most microplates, including deep-well microplates. Microplates can be stacked up to six plates high, to a maximum height of 127 mm. The unit's 12 suction cup feet and weight prevent the shaker from moving across the lab bench. Pulsing feature allows the user to program the unit to stop and start at a wide range of intervals. The repetitive stopping and starting action is ideal for difficult mixing applications, such as emulsification of highly viscous liquids or bi-polar liquids.
Ordering information: Supplied with top plate and tray pad set. Tray pad set and replacement top plate are available separately. Each unit is supplied complete with three plugs: EU, UK and CH for use throughout Europe.
Specifications for Product Family
Specifications
  • Catalog No:
  • 444-7016
  • VWRU14216-214
  • Model:
  • Microplate shaker, high speed
  • Microplate shaker, high speed
  • Speed:
  • 600 - 2500 min⁻¹ (programmable in 1 min⁻¹ increments)
  • 600 - 2500 min⁻¹ (programmable in 1 min⁻¹ increments)
  • Environmentally Preferable:
  • Waste Reducing
  • Speed accuracy:
  • ±2 %
  • ±2 %
  • Orbit:
  • 3,6 mm
  • 3,6 mm
  • Max. load:
  • 3,2 kg
  • 3,2 kg
  • Power:
  • 120 W
  • 120 W
  • Weight:
  • 22 kg
  • 22 kg
  • W×D×H:
  • 305×394×325 mm
  • 305×394×325 mm
Specifications
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • High speed microplate shaker
    1 * 1 items
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    444-7016
    Model
    Microplate shaker, high speed
    Speed
    600 - 2500 min⁻¹ (programmable in 1 min⁻¹ increments)
    Environmentally Preferable
    Waste Reducing
    Speed accuracy
    ±2 %
    Orbit
    3,6 mm
    Max. load
    3,2 kg
    Power
    120 W
    Weight
    22 kg
    W×D×H
    305×394×325 mm
      Waste Reducing
  • High speed microplate shaker
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU14216-214
    Model
    Microplate shaker, high speed
    Speed
    600 - 2500 min⁻¹ (programmable in 1 min⁻¹ increments)
    Environmentally Preferable
    Speed accuracy
    ±2 %
    Orbit
    3,6 mm
    Max. load
    3,2 kg
    Power
    120 W
    Weight
    22 kg
    W×D×H
    305×394×325 mm
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 444-0201
    Replacement tray
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 444-7014
    Tray pad set