Orders LinkContactUsLinkComponent
VWR®, サンプルディッパー
VWR®, サンプルディッパー
Supplier:   VWR Collection
VWR®, サンプルディッパー
Supplier:  VWR Collection
Certificates
VWR®, サンプルディッパー
VWR®, サンプルディッパー
5 Options Available Below
Product Details & Documents
These sample dippers feature a basic container with a screw-in PTFE encapsulated steel shaft.
  • Excellent chemical resistance, temperature resistant
  • The basic unit consists of a container with a single 600 mm shaft
  • Additional shafts (331-0005) can be screwed onto the handle to increase length
Suitable for use at high and low temperatures (–200 to +280 °C).
Product Family Options
Product Information
  • 説明
    Material
    長さ
    直径
    容量
    Availability
    Price
  • Sample dipper
    PTFE
    600 mm
    54 mm
    100 ml
    1 * 1 items
  • Sample dipper
    PTFE
    600 mm
    66 mm
    250 ml
    1 * 1 items
  • Sample dipper
    PTFE
    600 mm
    80 mm
    500 ml
    1 * 1 items
  • Sample dipper
    PTFE
    600 mm
    100 mm
    1000 ml
    1 * 1 items
Replacement shaft
Product Information
  • 説明
    Material
    長さ
    直径
    容量
    Availability
    Price
  • Replacement/extension shaft
    PTFE
    600 mm