Orders LinkContactUsLinkComponent
VWR®, サーモスタット用バス
Supplier:   VWR Collection
VWR®, サーモスタット用バス
Supplier:  VWR Collection
Certificates
VWR®, サーモスタット用バス
VWR®, サーモスタット用バス
7 Options Available Below
Product Details & Documents
Made from stainless steel or polycarbonate. Ideal for use with VWR® immersion thermostats. For use where the durability of stainless steel or the transparency of polycarbonate are advantageous.
  • Open baths, ideal for use with immersion thermostats from VWR® or other manufacturers
  • Available in stainless steel or polycarbonate
  • Robust and durable
  • Easy to clean
Note: The temperature ranges listed below apply only when the vessels are used with immersion thermostats; an immersion thermostat is required but not supplied with the bath.
Product Family Options
Polycarbonate
Product Information
  • 温度範囲
    容量
    Bath W×D×H
    W×D×H
    Availability
    Price
  • <85 °C
    8 L
    284×156×203 mm
    302×174×232 mm
    1 * 1 items
  • <85 °C
    14 L
    493×156×203 mm
    513×174×232 mm
    1 * 1 items
  • <85 °C
    17 L
    312×305×203 mm
    351×324×232 mm
    1 * 1 items
  • <85 °C
    23 L
    417×305×203 mm
    455×324×232 mm
    1 * 1 items
  • <85 °C
    28 L
    523×305×203 mm
    561×324×232 mm
    1 * 1 items
Stainless steel
Product Information
  • 温度範囲
    容量
    Bath W×D×H
    W×D×H
    Availability
    Price
  • <150 °C
    13 L
    229×165×203 mm
    321×289×244 mm
    1 * 1 items
  • <150 °C
    28 L
    419×216×203 mm
    530×327×244 mm