Orders LinkContactUsLinkComponent
VWR®, 特殊pH電極
VWR®, 特殊pH電極
VWR®, 特殊pH電極
Supplier:  VWR Collection
Certificates
VWR®, 特殊pH電極
VWR®, 特殊pH電極
5 Options Available Below
Product Family Options
Product Information
  • Type
    Temp.
    Material
    Electrolyte
    直径×長さ
    Connection
    For
    Availability
    Price
  • Flat extra wide, double junction (HDPE/Pellon)
    +10...+100 °C
    Epoxy
    Gel
    15×115 mm
    BNC
    Agar, microbiological samples, cheese, butter, water
    1 * 1 items
  • Low ionic, double juntion (Pellon)
    0...+100 °C
    Epoxy
    Gel
    12×150 mm
    BNC
    Low ionic strength solutions
    1 * 1 items
  • ORP (Redox), Pellon junction
    0...+100 °C
    Epoxy
    Gel
    12×150 mm
    BNC
    Electroplating baths
    1 * 1 items
  • pH only half cell
    0...+100 °C
    Epoxy
    12×150 mm
    BNC
    Water, creams, beverage
    1 * 1 items
  • pH only half cell
    0...+100 °C
    Glass
    12×150 mm
    DIN
    Water, creams, beverage