Orders LinkContactUsLinkComponent
クロマトグラフィーチューブ, PTFE
   Sustainable Options Available
Supplier:   HICHROM
クロマトグラフィーチューブ, PTFE
Supplier:  HICHROM
   Sustainable Options Available
Certificates
クロマトグラフィーチューブ, PTFE
12 Options Available Below
Product Details & Documents
PTFE
  • Flexible and economical
  • Excellent solvent resistance
Inert to all chemicals used in HPLC but suitable for low-pressure applications only. The most common tubing used to connect the solvent reservoir to the pump.
Specifications for Product Family
Specifications
  • Catalog No:
  • 16-PTFE-07-3M
  • 16-PTFE-07-5M
  • 16-PTFE-07-10M
  • 16-PTFE-20-3M
  • 16-PTFE-20-5M
  • 16-PTFE-20-10M
  • 16-PTFE-30-3M
  • 16-PTFE-30-5M
  • 16-PTFE-30-10M
  • 08-PTFE-62-3M
  • 08-PTFE-62-5M
  • 08-PTFE-62-10M
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Product Information
  • 内径
    外径
    長さ
    Availability
    Price
  • 0,18 mm
    1,59 mm
    3 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    16-PTFE-07-3M
    Environmentally Preferable
    Environmental certification
  • 0,18 mm
    1,59 mm
    5 m
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    16-PTFE-07-5M
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • 0,18 mm
    1,59 mm
    10 m
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    16-PTFE-07-10M
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • 0,50 mm
    1,59 mm
    3 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    16-PTFE-20-3M
    Environmentally Preferable
    Environmental certification
  • 0,50 mm
    1,59 mm
    5 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    16-PTFE-20-5M
    Environmentally Preferable
    Environmental certification
  • 0,50 mm
    1,59 mm
    10 m
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    16-PTFE-20-10M
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • 0,75 mm
    1,59 mm
    3 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    16-PTFE-30-3M
    Environmentally Preferable
    Environmental certification
  • 0,75 mm
    1,59 mm
    5 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    16-PTFE-30-5M
    Environmentally Preferable
    Environmental certification
  • 0,75 mm
    1,59 mm
    10 m
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    16-PTFE-30-10M
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • 1,58 mm
    3,18 mm
    3 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    08-PTFE-62-3M
    Environmentally Preferable
    Environmental certification
  • 1,58 mm
    3,18 mm
    5 m
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    08-PTFE-62-5M
    Environmentally Preferable
    Environmental certification
  • 1,58 mm
    3,18 mm
    10 m
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    08-PTFE-62-10M
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact