Orders LinkContactUsLinkComponent
クロマトグラフィーチューブ, PEEK
クロマトグラフィーチューブ, PEEK
Supplier:   HICHROM
クロマトグラフィーチューブ, PEEK
Supplier:  HICHROM
Certificates
クロマトグラフィーチューブ, PEEK
クロマトグラフィーチューブ, PEEK
26 Options Available Below
Product Details & Documents
PEEK
  • Inert and biocompatible
  • Excellent for high pressure applications (413 bar)
Ten times less oxygen permeable than PTFE and with a greater temperature resistance. Completely flexible, easy to cut and suitable for a wide range of applications. Can be used with PEEK fingertight fittings or stainless steel nuts and ferrules.
Product Family Options
Product Information
  • 説明
    内径
    外径
    長さ
    Availability
    Price
  • PEEK tubing, natural
    0,05 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, green stripe
    1,90 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, black stripe
    0,10 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, red stripe
    0,13 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, yellow stripe
    0,18 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, solid yellow
    0,18 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, blue stripe
    0,25 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, orange stripe
    0,50 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, green stripe
    0,76 mm
    1,58 mm
    3 m
    1 * 1 items
  • PEEK tubing, green stripe
    1,90 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, black stripe
    0,10 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, red stripe
    0,13 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, yellow stripe
    0,18 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, blue stripe
    0,25 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, orange stripe
    0,50 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, green stripe
    0,76 mm
    1,58 mm
    5 m
    1 * 1 items
  • PEEK tubing, natural
    0,05 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, green stripe
    1,90 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, black stripe
    0,10 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, red stripe
    0,13 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, yellow stripe
    0,18 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, solid yellow
    0,18 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, blue stripe
    0,25 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, orange stripe
    0,50 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, green stripe
    0,76 mm
    1,58 mm
    10 m
    1 * 1 items
  • PEEK tubing, orange stripe
    0,50 mm
    1,58 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m