Orders LinkContactUsLinkComponent
VWR®, チューブ、シリコーン
VWR®, チューブ、シリコーン
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, チューブ、シリコーン
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, チューブ、シリコーン
39 Options Available Below
Product Details & Documents
Silicone, translucent, peroxide cross-linked.
  • Good physical properties, pliant, elastic and flexible
  • Suitable for use with foodstuffs in accordance with FDA
  • Produced in compliance with GMP conditions, ISO 13485, certificate of conformity supplied with each roll
For laboratory and medical applications, pharmacies, biotechnology, fermentation and pharmaceutical production. Biologically neutral (USP Class VI). Compliant with Eur. Ph. (2nd ed.).
Temperature resistance: −50 to +200 °C (briefly up to 250 °C)
Hardness (Shore durometer A): 60 ±5
Density: 1,16
Specifications for Product Family
Specifications
  • Catalog No:
  • 228-0701
  • 228-1450
  • 228-1528
  • 228-0704
  • 228-1451
  • 228-0707
  • 228-1517
  • 228-1518
  • 228-0709
  • 228-0712
  • 228-1452
  • 228-1519
  • 228-0714
  • 228-1453
  • 228-1454
  • 228-4400
  • 228-0718
  • 228-0720
  • 228-0722
  • 228-1455
  • 228-1520
  • 228-1521
  • 228-1522
  • 228-1456
  • 228-0726
  • 228-0728
  • 228-1523
  • 228-1457
  • 228-0731
  • 228-1524
  • 228-1525
  • 228-1526
  • 228-1527
  • 228-1458
  • 228-1459
  • 228-1460
  • 228-1461
  • 228-1463
  • 228-1462
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
  • ISO 14001 (N° 2020/88219.3)
Specifications
Product Family Options
Product Information
  • チューブ内径
    壁の厚さ
    チューブ外径
    Availability
    Price
  • 1,00 mm
    1,00 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0701
    Environmentally Preferable
    Environmental certification
  • 1,50 mm
    0,75 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1450
    Environmentally Preferable
    Environmental certification
  • 1,60 mm
    1,60 mm
    4,80 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1528
    Environmentally Preferable
    Environmental certification
  • 2,00 mm
    1,00 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0704
    Environmentally Preferable
    Environmental certification
  • 2,00 mm
    2,00 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1451
    Environmentally Preferable
    Environmental certification
  • 3,00 mm
    1,00 mm
    5,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0707
    Environmentally Preferable
    Environmental certification
  • 3,00 mm
    1,50 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1517
    Environmentally Preferable
    Environmental certification
  • 3,00 mm
    2,00 mm
    7,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1518
    Environmentally Preferable
    Environmental certification
  • 4,00 mm
    1,00 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0709
    Environmentally Preferable
    Environmental certification
  • 4,00 mm
    2,00 mm
    8,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0712
    Environmentally Preferable
    Environmental certification
  • 4,00 mm
    1,50 mm
    7,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1452
    Environmentally Preferable
    Environmental certification
  • 4,00 mm
    3,00 mm
    10,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1519
    Environmentally Preferable
    Environmental certification
  • 5,00 mm
    1,50 mm
    8,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0714
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 5,00 mm
    2,00 mm
    9,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1453
    Environmentally Preferable
    Environmental certification
  • 5,00 mm
    2,50 mm
    10,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1454
    Environmentally Preferable
    Environmental certification
  • 5,00 mm
    3,00 mm
    11,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-4400
    Environmentally Preferable
    Environmental certification
  • 6,00 mm
    1,50 mm
    9,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0718
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 6,00 mm
    2,00 mm
    10,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0720
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 6,00 mm
    3,00 mm
    12,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0722
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 7,00 mm
    1,50 mm
    10,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1455
    Environmentally Preferable
    Environmental certification
  • 7,00 mm
    2,00 mm
    11,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1520
    Environmentally Preferable
    Environmental certification
  • 7,00 mm
    2,50 mm
    12,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1521
    Environmentally Preferable
    Environmental certification
  • 7,00 mm
    3,00 mm
    13,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1522
    Environmentally Preferable
    Environmental certification
  • 8,00 mm
    1,50 mm
    11,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-1456
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 8,00 mm
    2,00 mm
    12,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0726
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 8,00 mm
    3,00 mm
    14,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-0728
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 8,00 mm
    4,00 mm
    16,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1523
    Environmentally Preferable
    Environmental certification
  • 9,00 mm
    2,00 mm
    13,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1457
    Environmentally Preferable
    Environmental certification
  • 10,00 mm
    2,00 mm
    14,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-0731
    Environmentally Preferable
    Environmental certification
  • 10,00 mm
    3,00 mm
    16,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1524
    Environmentally Preferable
    Environmental certification
  • 10,00 mm
    4,00 mm
    18,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-1525
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 12,00 mm
    1,75 mm
    15,50 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1526
    Environmentally Preferable
    Environmental certification
  • 12,00 mm
    2,50 mm
    17,00 mm
    Log in for availability
    Log in for availability
    1 * 25 m
    1 * 25 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1527
    Environmentally Preferable
    Environmental certification
  • 15,00 mm
    3,00 mm
    21,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    228-1458
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (N° 2020/88219.3)
      Low Manufacturing Impact
  • 18,00 mm
    3,00 mm
    24,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1459
    Environmentally Preferable
    Environmental certification
  • 20,00 mm
    3,50 mm
    27,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1460
    Environmentally Preferable
    Environmental certification
  • 25,00 mm
    5,00 mm
    35,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1461
    Environmentally Preferable
    Environmental certification
  • 40,00 mm
    5,00 mm
    50,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1463
    Environmentally Preferable
    Environmental certification
  • 30,00 mm
    5,00 mm
    40,00 mm
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m
    Specifications:
    More
    Specifications
    Cat. No.
    228-1462
    Environmentally Preferable
    Environmental certification