VWR®, クロージャ
LDPE, 212-1809/212-1837, Natural, 10 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
These closures are suitable for test and culture tubes.
- Flexible, high mechanical resistance
Closures are made from LDPE.
Documents
Specifications
- キャップサイズ:10 mm
- For tubes:212-1809/212-1837
- Colour:Natural
- Material:LDPE
Complementary Products
Product Information
- Catalog #容量長さ外径Material内径AvailabilityPrice