Orders LinkContactUsLinkComponent
VWR®, 汎用オーブン温度計
Supplier:   VWR Collection
VWR®, 汎用オーブン温度計
Supplier:  VWR Collection
Certificates
VWR®, 汎用オーブン温度計
VWR®, 汎用オーブン温度計
5 Options Available Below
Product Details & Documents
These oven thermometers contain red organic liquid against white back glass.
  • Partial immersion thermometers feature a round glass bulge in the body to hold them in place
  • PTFE coating helps prevent liquid and glass contamination if the thermometer breaks
  • Individually serialised
Product Family Options
Standard coating
Product Information
  • Range
    Accuracy
    Division
    長さ
    Immersion depth
    Availability
    Price
  • 0…+200 °C
    ±2,0; ±2,5 (>100 °C)
    1,0 °C
    280 mm
    76 mm
    1 * 1 items
  • +20…+130 °C
    ±2,0 °C
    2,0 °C
    135 mm
    Total
    1 * 1 items
PTFE coating
Product Information
  • Range
    Accuracy
    Division
    長さ
    Immersion depth
    Availability
    Price
  • 0…+200 °C
    ±2,0; ±2,5 (>100 °C)
    1,0 °C
    280 mm
    76 mm
    1 * 1 items
  • 0…+200 °C
    ±2,0; ±2,5 (>100 °C)
    1,0 °C
    405 mm
    125 mm
    1 * 1 items
  • +20…+130 °C
    ±2,0 °C
    2,0 °C
    135 mm
    Total