Orders LinkContactUsLinkComponent
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途
Supplier:  VWR Collection
Certificates
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途
10 Options Available Below
Product Details & Documents
These multi-purpose vinyl tapes, wound on a plastic core, are ideal for use in cleanroom maintenance and support areas.
  • Ideal for colour coding, sealing, aisle marking and batching
  • Matt finish can be written on with a cleanroom pen
Length: 32,9 m.
Different colours available.
 : Single-packed in an easy tear, perforated bag.
Product Family Options
Product Information
  • 説明
    Colour
    Availability
    Price
  • Cleanroom tape, general purpose
    White
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    White
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Yellow
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Transparent
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 36 Roll
    1 * 36 Roll
  • Cleanroom tape, general purpose
    Dark blue
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Dark green
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Light blue
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Light green
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Orange
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Cleanroom tape, general purpose
    Red
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll