Orders LinkContactUsLinkComponent
VWR®, ベンチプロテクタ
VWR®, ベンチプロテクタ
VWR®, ベンチプロテクタ
Supplier:  VWR Collection
Certificates
VWR®, ベンチプロテクタ
VWR®, ベンチプロテクタ
2 Options Available Below
Product Details & Documents
High quality, smooth, absorbent paper that quickly absorbs liquid spills with a thick, laminated polyethylene layer that prevents seeping onto the work surface.
  • Retains leaked reagents in radiochemical laboratories preventing radioactive contamination
  • Enables recovery of expensive leaked materials
  • Breakage protection for hard surfaces
  • Absorption layer for removing water or solvent from desiccant chambers
  • Impermeable protective material for lining animal cages
Absorbance rate: 115-9220 and 115-9221: 500 ml/m²
Absorbance rate: 115-0673: 750 ml/m²
Product Family Options
Product Information
  • 説明
    Colour
    Packed
    長さ×幅
    Availability
    Price
  • Absorption: Approx. 500 ml/m²
    White
    Single sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • Absorption: Approx. 500 ml/m²
    White
    Roll
    460×50 000 mm