Orders LinkContactUsLinkComponent
VWR®, 攪拌棒
VWR®, 攪拌棒
Supplier:   VWR Collection
VWR®, 攪拌棒
Supplier:  VWR Collection
Certificates
VWR®, 攪拌棒
VWR®, 攪拌棒
3 Options Available Below
Product Details & Documents
Solid PTFE.
  • Inert and will not scratch, can be used up to 280 °C
Product Family Options
Product Information
  • 説明
    Material
    直径
    長さ
    Availability
    Price
  • Stirring rods, PTFE
    PTFE
    8 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Stirring rods, PTFE
    PTFE
    8 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Stirring rods, PTFE
    PTFE
    8 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items