Orders LinkContactUsLinkComponent
VWR®, マグネット式羽根型スターラー、マイクロ
VWR®, マグネット式羽根型スターラー、マイクロ
Supplier:   VWR Collection
VWR®, マグネット式羽根型スターラー、マイクロ
Supplier:  VWR Collection
Certificates
VWR®, マグネット式羽根型スターラー、マイクロ
VWR®, マグネット式羽根型スターラー、マイクロ
5 Options Available Below
Product Details & Documents
PTFE covered, Alnico V magnetic core. Smooth surface. For stirring in test tubes, centrifuge tubes and micro vials. The wings can also be trimmed to size if needed.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Documents
Product Family Options
Triangular
Product Information
  • 高さ
    直径
    For
    Availability
    Price
  • 9 mm
    5,5 mm
    Container with max. Ø 6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 11 mm
    8 mm
    Container with max. Ø 9 - 10 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 16 mm
    10 mm
    Container with max. Ø 11 - 12 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
Semicircular
Product Information
  • 高さ
    直径
    For
    Availability
    Price
  • 10 mm
    13 mm
    Container with max. Ø 14 - 15 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 20 mm
    13 mm
    Container with max. Ø 14 - 15 mm