VWR®, マグネティックスターリングディスク
Supplier: VWR Collection
Jump to:
About this item
PTFE covered, strong Alnico V magnetic core. The special shape with raised surface in the centre ensures very good agitation even at low speeds. Ideal for use in glass beakers.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
3 Options Available Below
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. The special shape with raised surface in the centre ensures very good agitation even at low speeds. Ideal for use in glass beakers.
- Excellent chemical resistance
- Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Documents
442-0492
Images 360
Product Family Options
Product Information
- 長さ直径AvailabilityPrice