Orders LinkContactUsLinkComponent
VWR®, マグネット式スターラーバー、プレーン
VWR®, マグネット式スターラーバー、プレーン
VWR®, マグネット式スターラーバー、プレーン
Supplier:  VWR Collection
Certificates
VWR®, マグネット式スターラーバー、プレーン
VWR®, マグネット式スターラーバー、プレーン
19 Options Available Below
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Product Family Options
Product Information
  • 長さ
    直径
    Availability
    Price
  • 10 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0262
  • 13 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0263
  • 15 mm
    4,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0264
  • 20 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0265
  • 20 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0266
  • 25 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0267
  • 25 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0268
  • 30 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0269
  • 35 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0270
  • 40 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0271
  • 50 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0272
  • 57 mm
    27 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0278
  • 60 mm
    7 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0273
  • 65 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0274
  • 70 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0275
  • 75 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0276
  • 80 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0277
  • 108 mm
    27 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0279
  • 159 mm
    27 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
    More
    Specifications
    Cat. No.
    442-0280