Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, rectangular
Supplier:  VWR Collection
Certificates
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular
4 Options Available Below
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Product Family Options
Product Information
  • 長さ
    高さ
    Availability
    Price
  • 12 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 25 mm
    5,5 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 35 mm
    6 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 50 mm
    7,5 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items