Orders LinkContactUsLinkComponent
VWR®, 実験用マーカー
VWR®, 実験用マーカー
VWR®, 実験用マーカー
VWR®, 実験用マーカー
These versatile laboratory markers are designed to write on wet or dry surfaces, and are suitable for industrial, biomedical and educational laboratories.
  • Durable tip and generous ink supply ensure a long product life
  • Ink dries instantly, allowing immediate handling
  • Taper tip from 4mm to 1mm
    Alcohol-resistant
  • Non sterile
  • Made without natural rubber latex
These markers provide permanent marking on plastic disposables. When used on glass, metal or porcelain surfaces, the ink will not smear when subjected to water, and can be washed off these surfaces using various solvents, allowing repeated marking.
Product Family Options
  • 説明
    Colour
  • Laboratory markers, fine tip
    Black
    1 * 10 items
    1 * 10 items
  • Laboratory markers, fine tip
    Green
    1 * 10 items
    1 * 10 items
  • Laboratory markers, fine tip
    Blue
    1 * 10 items
    1 * 10 items
  • Laboratory markers, fine tip
    Red