Orders LinkContactUsLinkComponent
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
   Sustainable Options Available
Supplier:   VWR Collection
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
18 Options Available Below
Product Details & Documents
Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
  • High tensile strength, and excellent heat resistance, for enhanced copier and printing efficiency
  • PE-binding technology results in very low levels of particles, non volatile residue and ion extractables
  • Unique precision-cutting technology guarantees consistent sizing and cleanliness
  • Paper is web-cleaned on both sides prior to sheeting
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double-bagged in an ISO Class 5 (FED-STD-209E Class 100) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10) to ISO Class 6 (FED-STD-209E Class 1000) cleanrooms.
Poly paper excels in printability. Synthetic paper excels in tensile strength.
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
Packaging: Paper is packed 250 sheets per pack, 10 packs per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 115-0149
  • 115-0150
  • 115-0156
  • 115-1961
  • 115-1952
  • 115-1958
  • 115-1954
  • 115-1959
  • 115-0163
  • 115-0164
  • 115-0165
  • 115-0166
  • 115-1955
  • 115-1956
  • 115-1957
  • 115-1951
  • 115-1960
  • 115-1953
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
Product Family Options
Product Information
  • 説明
    Colour
    Weight
    Availability
    Price
  • Latex-free paper, A4 size, poly substrate
    White
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0149
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, poly substrate
    Blue
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0150
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, poly substrate
    White
    105 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0156
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A2 size, synthetic substrate
    White
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1961
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A3 size, synthetic substrate
    Blue
    85 g/m²
    Log in for availability
    Log in for availability
    1 * 1.250 items
    1 * 1.250 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1952
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A3 size, synthetic substrate
    White
    85 g/m²
    Log in for availability
    Log in for availability
    1 * 1.250 items
    1 * 1.250 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1958
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A3 size, synthetic substrate
    Blue
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 1.250 items
    1 * 1.250 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1954
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A3 size, synthetic substrate
    White
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 1.250 items
    1 * 1.250 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1959
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    White
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0163
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    Blue
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0164
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    Yellow
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0165
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    Green
    80 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-0166
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    Pink
    85 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1955
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    White
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1956
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    White
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1957
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A4 size, synthetic substrate
    Blue
    105 g/m²
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1951
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A5 size, synthetic substrate
    White
    104 g/m²
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1960
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Latex-free paper, A5 size, synthetic substrate
    Blue
    105 g/m²
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    115-1953
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging