Orders LinkContactUsLinkComponent
VWR®, 実験記録簿
VWR®, 実験記録簿
Supplier:   VWR Collection
VWR®, 実験記録簿
Supplier:  VWR Collection
Certificates
VWR®, 実験記録簿
VWR®, 実験記録簿
6 Options Available Below
Product Details & Documents
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm). Used to record and log data relating to laboratory equipment, temperature, balance calibration, reagents, animal maintenance and sample receipts.
  • All pages for logging documentation have specific column headings for easy data entry
  • Pages sewn in sections, books have round binding so that they lay flat
  • Waterproof and tough
All books made in accordance with GLP, GMP and GCP guidelines.
Certifications: GLP, GMP and GCP guidelines.
Product Family Options
Product Information
  • 説明
    Colour
    No. of pages
    Availability
    Price
  • Log book, laboratory equipment
    Red
    96
    1 * 1 items
  • Log book, animal maintenance
    Black
    168
    1 * 1 items
  • Log book, balance calibration
    Green
    168
    1 * 1 items
  • Log book, reagents
    Dark red
    168
    1 * 1 items
  • Log book, sample receipt
    Red
    168
    1 * 1 items
  • Log book, temperature
    White
    168