Orders LinkContactUsLinkComponent
VWR®, 実験ノート
Supplier:   VWR Collection
VWR®, 実験ノート
Supplier:  VWR Collection
Certificates
VWR®, 実験ノート
VWR®, 実験ノート
25 Options Available Below
Product Details & Documents
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive safe paper (90 gsm). Includes laboratory notebook guidelines page, table of contents and user data page. Available in different language formats: English only, English-French, English-German, English-Spanish and English-Italian.
  • Pages are sewn in sections with round binding so that book lays flat
  • Scientific 5 mm grid or ruled format with date and witness signature blocks
  • Waterproof
Format: A4 (21×29,7 cm) or A5 (14,8×21 cm). A4 books available in three sizes: 72, 120 and 216 pages. A5 available with 96 pages.
Colour: blue
All books made in accordance with GLP, GMP and GCP guidelines.
Product Family Options
Product Information
  • 説明
    Colour
    Type
    Version
    Paper size
    No. of pages
    Availability
    Price
  • Lab book
    Blue
    Ruled
    English only
    A4
    216
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English only
    A5
    96
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-French
    A4
    72
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-French
    A4
    120
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-French
    A4
    216
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-French
    A5
    96
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-French
    A4
    72
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-French
    A4
    120
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-French
    A4
    216
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-French
    A5
    96
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-German
    A4
    216
    1 * 1 items
  • Lab book
    Blue
    Grid
    English-German
    A5
    96
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-German
    A4
    72
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English-German
    A4
    120
    1 * 1 items
  • Lab book
    Blue
    Grid
    English only
    A4
    72
    1 * 1 items
  • Lab book
    Blue
    Grid
    English only
    A4
    120
    1 * 1 items
  • Lab book
    Blue
    Grid
    English only
    A4
    216
    1 * 1 items
  • Lab book
    Blue
    Grid
    English only
    A5
    96
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English only
    A4
    72
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English only
    A4
    120
    1 * 1 items
  • Lab book
    Blue
    Ruled
    English
    A4
    216
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $132.48
    $132.48
    1 * 1 items
       $132.48
  • Lab book
    Blue
    Ruled
    English
    A4
    120
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Lab book
    Blue
    Grid
    English
    A4
    216
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $132.48
    $132.48
    1 * 1 items
       $132.48
  • Lab book
    Blue
    Grid
    EN/FR/IT/SP/DE
    225×345 mm
    144
    1 * 1 items
  • Lab book
    Blue
    Ruled
    EN/FR/IT/SP/DE
    225×345 mm
    144