VWR®, シーリングマット、PCRプレート用
Sealing mats, silicone, round wells
Catalog # 732-0589
Supplier: VWR Collection
VWR®, シーリングマット、PCRプレート用
Catalog # 732-0589
Supplier: VWR Collection
Jump to:
Product Details & Documents
Transparent silicone mats fit most brands of 96-well PCR plates.
- Sustainable design based on virgin resins
- Can be cleaned and reused
- Autoclavable at 121 °C
Designed to reduce evaporation when cycling 96-well plates.
Specifications
- 説明:Sealing mats, silicone, round wells
- Seq:12
- Cat. No.:732-0589
- Environmentally Preferable:Low Manufacturing Impact|Sustainable Packaging