Orders LinkContactUsLinkComponent
VWR®, マイクロチューブラック、フローティング
VWR®, マイクロチューブラック、フローティング
VWR®, マイクロチューブラック、フローティング
VWR®, マイクロチューブラック、フローティング
PP.
  • Removable feet enable microtubes to be removed quickly and easily
  • Also suitable for cryotubes with up to 2 ml capacity
  • Stable, good chemical resistance
For maintaining microtubes with capacities from 0,5 to 2 ml at constant temperature in water baths. Racks float even when full. The tubes are held firmly in place in the frame and do not float. Handles are provided for easier insertion and removal from the water bath and for carrying. Not autoclavable.
Product Family Options
  • 穴の直径
    Colour
    穴の数
    For tubes
    W×D×H
    Form
  • 98 mm
    White
    20
    1,5/2,0 ml
    Round
    1 * 4 items
    1 * 4 items
  • White
    16 (4×4)
    1,5/2,0 ml
    102×102×60 mm
    Square
    1 * 4 items
    1 * 4 items