Orders LinkContactUsLinkComponent
VWR®, ラック、エッペンチューブ用
VWR®, ラック、エッペンチューブ用
VWR®, ラック、エッペンチューブ用
VWR®, ラック、エッペンチューブ用
PP.
  • For freezer use
  • Side handles for secure handling
  • Numbered for easy sample identification
Space for up to 80×1,5 or 2,0 ml microcentrifuge tubes. 5×16 array with conical-shaped holes to hold microtubes securely.
 : The assorted colour set includes one rack each in blue, green, pink, yellow and orange.
Product Family Options
  • Colour
    穴の数
    W×D×H
  • Natural
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Orange
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Yellow
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Blue
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Green
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Pink
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items
  • Assorted colours
    80 (5×16)
    225×67×29 mm
    1 * 5 items
    1 * 5 items