Orders LinkContactUsLinkComponent
VWR®, ラック、エッペンチューブ用
VWR®, ラック、エッペンチューブ用
Supplier:   VWR Collection
VWR®, ラック、エッペンチューブ用
Supplier:  VWR Collection
Certificates
VWR®, ラック、エッペンチューブ用
VWR®, ラック、エッペンチューブ用
6 Options Available Below
Product Details & Documents
PP.
  • For freezer use
  • Side handles for secure handling
  • Numbered for easy sample identification
For 96 (8×12) microtubes with 0,5 ml capacity or 96 microtubes with 1,5/2 ml capacity. Conical, tapered openings to hold tubes securely. Complete with snap-on lid, which can also be used as a protective base.
Ordering information: The assorted colour kit includes one rack each coloured blue, green, pink, yellow and orange.
Product Family Options
Product Information
  • Colour
    穴の数
    W×D×H
    Availability
    Price
  • Natural
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Yellow
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Blue
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Green
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Pink
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Assorted colours
    96 (8×12)
    246×121×51 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items