VWR®, ラック、凍結ボックス用
Supplier: VWR Collection
Jump to:
About this item
Stainless steel, with handles.
Horizontal racks fit cryoboxes up to 133×133 mm.
2 Options Available Below
Product Details & Documents
Stainless steel, with handles.
Horizontal racks fit cryoboxes up to 133×133 mm.
Documents
VWR® Collection Selection Catalogue VWR® Lab bric-a-brac