Orders LinkContactUsLinkComponent
VWR®, ディスペンサーチップ、クラシック
VWR®, ディスペンサーチップ、クラシック
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, ディスペンサーチップ、クラシック
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, ディスペンサーチップ、クラシック
VWR®, ディスペンサーチップ、クラシック
8 Options Available Below
Product Details & Documents
By using the direct displacement method, these tips work well for infectious and volatile liquids, as the fluid never will get in touch with the instrument which avoids any contamination.
  • Compatible with: VWR Stepper and Stepper Pro, Ritter® Ripette® and Ripette® pro, Brand® HandyStep® and HandyStep® electronic*, Eppendorf® Multipette® 4780, Gilson Repetman® and Easystep (* without automatic tip recognition)
  • Sustainable design based on virgin resins
  • Eight different tip sizes from 0,05 to 50 ml
  • Sterile tips guaranteed free of DNA, DNase, RNase, ATP and pyrogens
NOTE: The 25 ml and 50 ml tips require an adapter, which needs to be ordered separately. When using 0,05 ml tips it has to be taken into account to attach a 10 µl manual pipette tip accordingly.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-1001
  • 613-1011
  • 613-1004
  • 613-1012
  • 613-1005
  • 613-1013
  • 613-1006
  • 613-1007
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Specifications
Product Family Options
Product Information
  • 容量
    Sterile
    Packed
    Availability
    Price
  • 0,5 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-1001
    Environmentally Preferable
    Environmental certification
  • 2,5 ml
    +
    Individual blister
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1011
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 5,0 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1004
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 5,0 ml
    +
    Individual blister
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1012
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 12,5 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1005
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 12,5 ml
    +
    Individual blister
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1013
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 25,0 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1006
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 50,0 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1007
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Adapter
Product Information
  • Catalog #
    説明
    Availability
    Price
  • Adapter for 25 ml tips
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $0.00