VWR®, ストローピペット、英語版のみ
Straw pipettes, 1 ml, 25 per bag
Supplier: VWR Collection
Jump to:
Product Details & Documents
Straw pipettes made from PP, irradiated, disposable, sterile, without graduation.
- Traceability including the expiry date on the secondary packaging
- Zip bag can be pressed closed ensuring product protection
- Tamper-evident seal guarantees the sterility of the product
Caution: *The product(s) below are outside the scope of the EU Directive 2019/904
Documents
Specifications
- Packed:25 per bag
- 容量:1 ml
- 説明:Straw pipettes
- Seq:01