VWR®, 安全バルブピペットフィラー
Safety bulb pipette fillers, 15 ml, Red/white
Supplier: VWR Collection
Jump to:
Product Details & Documents
Red rubber with white plastic connector and silicone conical adapter, autoclavable.
- For large and small capacity pipettes
- Conical adapter connects pipettes more securely, avoids breakages because it requires less force to fit on the bulb
- Elastic cord for attaching the bulb to a reagent vessel or to a user's wrist
Capacity: 15 ml
Documents
CoC Avantor VWR® Safety Bulb Pipette Fillers.pdf
Conformity declaration
Specifications
- Colour:Red/white
- 説明:Safety bulb pipette fillers, 15 ml
- Seq:01