Orders LinkContactUsLinkComponent
VWR®, ピペットバルブ
VWR®, ピペットバルブ
Supplier:   VWR Collection
VWR®, ピペットバルブ
Supplier:  VWR Collection
Certificates
VWR®, ピペットバルブ
VWR®, ピペットバルブ
2 Options Available Below
Product Details & Documents
Natural latex.
  • For Pasteur pipettes, small pipettes and medical drop counters
Capacity: 1 to 2 ml
Length: 43/52 mm
Product Family Options
Product Information
  • 説明
    容量
    Colour
    Availability
    Price
  • Dropper bulbs, 43 mm
    1 ml
    Natural
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items
  • Dropper bulbs, 52 mm
    2 ml
    Natural
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items