Orders LinkContactUsLinkComponent
VWR®, パスツールピペット
VWR®, パスツールピペット
VWR®, パスツールピペット
Supplier:  VWR Collection
Certificates
VWR®, パスツールピペット
VWR®, パスツールピペット
9 Options Available Below
Product Details & Documents
Soda-lime glass, disposable.
  • Non sterile
  • Automated manufacturing process ensures total production uniformity
  • Tapered upper opening for cotton ball plugging
  • Top body outer diameter: 7,1 mm, wall thickness: 0,53 mm, jet outer Ø: 1,5 mm
Product Family Options
Product Information
  • 説明
    容量
    長さ
    Availability
    Price
  • Non plugged
    2 ml
    150 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Non plugged
    2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Non plugged
    2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $124.03
    $124.03
    1 * 1.000 items
       $124.03
  • Non plugged, sealed tip, breakable
    2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Non plugged, sealed tip, breakable
    2 ml
    270 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Pre-plugged
    2 ml
    150 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Pre-plugged
    2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Pre-plugged, sealed tip, breakable
    2 ml
    230 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • Pre-plugged, sealed tip, breakable
    2 ml
    270 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items