VWR®, セルフ・ウインギング・モッピングシステム
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
- Autoclavable; resistant to gamma rays and ETO
- Resistant to the most widely available disinfectants and detergents
- Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
- Replacement foam mop heads are available with stainless steel or galvanised steel clips
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
- Autoclavable; resistant to gamma rays and ETO
- Resistant to the most widely available disinfectants and detergents
- Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
- Replacement foam mop heads are available with stainless steel or galvanised steel clips
Product Family Options
Replacement sponges
- 説明
Sponges (B)
- 説明