Orders LinkContactUsLinkComponent
VWR®, モップ、平型、クリーンルーム
VWR®, モップ、平型、クリーンルーム
VWR®, モップ、平型、クリーンルーム
Supplier:  VWR Collection
Certificates
VWR®, モップ、平型、クリーンルーム
VWR®, モップ、平型、クリーンルーム
5 Options Available Below
Product Details & Documents
Foam with 100% polyester cover. For applying disinfectants during pharmaceutical production or for aseptic processes. The flat mop head is ideally suited for cleaning large surfaces such as floors, walls and ceilings. Low profile, thick padding and angled head for ease of use.
  • Autoclavable, washable and reusable
  • Good moisture bonding provides optimum contact with the disinfectant
  • Available in sterile and non sterile versions
  • Resistant to strong disinfectants
Suitable for Class 10, ISO 4, M2 cleanrooms.
Ordering information: The complete mop consists of the mop head, a matching stainless steel adapter, and the mop handle. Please select the required components and order each separately.
Product Family Options
Product Information
  • 説明
    寸法
    Availability
    Price
  • Mop head, non sterile
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
  • Mop head, non sterile
    356 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
  • Mop head, non sterile
    457 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items - $965.05
    $965.05
    1 * 24 items
       $965.05
  • Mop head, sterile
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
  • Mop head, sterile
    356 mm
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items