VWR®, スライドホルダーキャビネット
Microscope slide holders, White, Cardboard, 125×75×1 mm, 400
631-0823
Cardboard microscope slide holders offer instant reference and fast, fingertip retrieval.
- Cardboard fitted with a protective plastic frame, which provides secure protection of specimens
- Each slide holder card accepts four slides of any thickness
- Space for identification and comments
Cards can be added, removed or changed for fast updating.
- No. of slides:400
- Colour:White
- 説明:Microscope slide holders
- Material:Cardboard
- Seq:02
- W×D×H:125×75×1 mm