VWR®, 顕微鏡スライド
- Clear, white glass
- Cut or bevelled edges
- Available plain or frosted
W×D×H: 76×26×1 mm
- Clear, white glass
- Cut or bevelled edges
- Available plain or frosted
W×D×H: 76×26×1 mm
Documents
Product Family Options
- 説明ColourMaterial厚さ長さ×幅
- Slide with bevelled edges and plain endTransparentFloat glass1,0 mm76×26 mm1 * 50 items
- Slide with bevelled edges and frosted endTransparentFloat glass1,0 mm76×26 mm1 * 50 items