Orders LinkContactUsLinkComponent
VWR®, ラボラトリージャッキ用アクセサリー
VWR®, ラボラトリージャッキ用アクセサリー
VWR®, ラボラトリージャッキ用アクセサリー
VWR®, ラボラトリージャッキ用アクセサリー
Extension panels: Aluminium, grey/black anodised. Increase the size of the work surface.
Stand plates: Aluminium, anodised grey/black. For securing to support stands.
Stand rods: Aluminium, anodised yellow. Can only be used in conjunction with the two extension panels 245-0017, 650 mm long, 12 mm diameter.
Stand fixture: Aluminium, anodised. For fixing laboratory racks to stand rods, etc.
Aluminium, anodised yellow. Can only be used in conjunction with the two extension panels 245-0017, 650 mm long, 12 mm diameter.
Product Family Options
  • 説明
    For
    Base
  • Support
    245-0005, 245-0006, 245-0007, 245-0008
    -
    1 * 1 items
    1 * 1 items
  • Support rod
    245-0006, 245-0007, 245-0008
    -
    1 * 1 items
    1 * 1 items
  • Retort stand plate
    Sockets up to 16 mm
    200×200 mm
    1 * 1 items
    1 * 1 items
  • Additional plate
    245-0006, 245-0007, 245-0008
    180×260 mm
    1 * 1 items
    1 * 1 items
  • Additional plate
    245-0006, 245-0007, 245-0008
    200×200 mm
    1 * 1 items
    1 * 1 items
  • Additional plate
    245-0006, 245-0007, 245-0008
    300×300 mm
    1 * 1 items
    1 * 1 items
  • Additional plate
    245-0006, 245-0007, 245-0008
    320×220 mm
    1 * 1 items
    1 * 1 items
  • Additional plate
    245-0009
    400×500 mm