Orders LinkContactUsLinkComponent
VWR®, 遠心分離フィルター
VWR®, 遠心分離フィルター
VWR®, 遠心分離フィルター
VWR®, 遠心分離フィルター
PP insert and receiver tube. MWCO filters are ideal to concentrate, purify and desalt peptides, proteins, oligonucleotides, DNA and RNA.
  • Low binding, typical recovery greater than 90%
  • Sealed membrane
  • Fit rotors that accept 1,5 ml tubes
These 0,2 and 0,45 µm centrifugal filters are used for particulate removal, sample clarification or pre-filtration clean-up of large agarose or acrylamide gel debris prior to further purification with a MWCO device.
Fits rotors that accept 1,5 ml tubes
  • Catalog No:
  • 516-0227
  • 516-0228
  • 516-0229
  • 516-0230
  • 516-0231
  • 516-0232
  • 516-0233
  • 516-0234
  • 516-0235
  • 516-0236
  • Filtration area:
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • 0,28 cm²
  • Max. RCF:
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • 14000 ×g
  • Dead volume:
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • <5 µl
  • Sample volume:
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Max. 500 µl
  • Temperature:
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
  • 0...40 °C
Product Family Options
PES (polyethersulfone) membrane
  • MWCO
    細孔サイズ
    Colour code
  • 3 kD
    Grey
    1 * 100 items
    1 * 100 items
    Specifications
    Cat. No.
    516-0227
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 3 kD
    Grey
    1 * 500 items
    1 * 500 items
    Specifications
    Cat. No.
    516-0228
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 10 kD
    Blue
    1 * 100 items
    1 * 100 items
    Specifications
    Cat. No.
    516-0229
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 10 kD
    Blue
    1 * 500 items
    1 * 500 items
    Specifications
    Cat. No.
    516-0230
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 30 kD
    Red
    1 * 100 items
    1 * 100 items
    Specifications
    Cat. No.
    516-0231
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 30 kD
    Red
    1 * 500 items
    1 * 500 items
    Specifications
    Cat. No.
    516-0232
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
Nylon membrane
  • MWCO
    細孔サイズ
    Colour code
  • 0,20 µm
    Turquoise
    1 * 100 items
    1 * 100 items
    Specifications
    Cat. No.
    516-0233
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 0,20 µm
    Turquoise
    1 * 500 items
    1 * 500 items
    Specifications
    Cat. No.
    516-0234
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 0,45 µm
    Purple
    1 * 100 items
    1 * 100 items
    Specifications
    Cat. No.
    516-0235
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C
  • 0,45
    Purple
    1 * 500 items
    1 * 500 items
    Specifications
    Cat. No.
    516-0236
    Filtration area
    0,28 cm²
    Max. RCF
    14000 ×g
    Dead volume
    <5 µl
    Sample volume
    Max. 500 µl
    Temperature
    0...40 °C