Orders LinkContactUsLinkComponent
VWR®, 使い捨て秤量皿
Supplier:   VWR Collection
VWR®, 使い捨て秤量皿
Supplier:  VWR Collection
Certificates
VWR®, 使い捨て秤量皿
VWR®, 使い捨て秤量皿
5 Options Available Below
Product Details & Documents
Aluminium, light gauge.
  • Flanged edge with curled lip for increased stability
  • Contaminant- and oil-free
  • Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
Product Family Options
Product Information
  • Int.Ø×H
    Availability
    Price
  • 44×13 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 63×18 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 65×30 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 110×29 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • 180×25 mm