Avantor® Hichrom C8, HPLCカラム
These Avantor® Hichrom C8 columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra pure silica, advanced bonding technology, superior column specification and comprehensive batch validation all contribute to the columns’ excellent reproducibility.
- Ultra pure, base deactivated porous silica
- Suitable for acidic, basic and neutral molecules
- Particle sizes 3,5 and 5 μm available
Combining ultra pure silica with advanced bonding technology results in a densely bonded silica that is both highly robust and highly reproducible. Unpredictable interactions between residual silanol sites on the silica surface and the analyte are effectively eliminated. Comprehensive batch validation ensures absolute batch to batch reproducibility is maintained with acidic, basic and neutral molecules.
Guard cartridges are available for C8 column dimensions, to further extend column lifetime by preventing irreversible adsorption and frit blockage at the top of the column.
: Other dimensions columns and guard cartridges are available on request.
: Supplied with a test chromatogram and batch validation certificate.
These Avantor® Hichrom C8 columns offer high performance in order to tackle the most challenging reversed-phase applications. The use of ultra pure silica, advanced bonding technology, superior column specification and comprehensive batch validation all contribute to the columns’ excellent reproducibility.
- Ultra pure, base deactivated porous silica
- Suitable for acidic, basic and neutral molecules
- Particle sizes 3,5 and 5 μm available
Combining ultra pure silica with advanced bonding technology results in a densely bonded silica that is both highly robust and highly reproducible. Unpredictable interactions between residual silanol sites on the silica surface and the analyte are effectively eliminated. Comprehensive batch validation ensures absolute batch to batch reproducibility is maintained with acidic, basic and neutral molecules.
Guard cartridges are available for C8 column dimensions, to further extend column lifetime by preventing irreversible adsorption and frit blockage at the top of the column.
: Other dimensions columns and guard cartridges are available on request.
: Supplied with a test chromatogram and batch validation certificate.
- Catalog No:
- HI-3.5C8-100AF
- HI-3.5C8-50A
- HI-3.5C8-75A
- HI-3.5C8-100A
- HI-3.5C8-150A
- HI-5C8-150AM
- HI-5C8-50AS
- HI-5C8-150AE
- HI-5C8-150AF
- HI-5C8-250AF
- HI-5C8-150A
- HI-5C8-250A
- HI-3.5C8-10C5
- HI-5C8-10C5
- HICHHI-5C8-10CP
- HI-5C8-10CP3
- HI-5C8-100P
- HI-5C8-150P
- HI-5C8-250P
- HI-5C8-50P
- HI-5C8-50SP1
- HI-5C8-75P
- HI-5C8-100P3
- HI-5C8-150P3
- HI-5C8-150SP1
- HI-5C8-250P3
- HI-5C8-50P3
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- 250 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Product Family Options
- 説明カラム形式Phase細孔サイズ粒子サイズ内径長さ
Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-100AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-50AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-75AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-100AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-150AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-150AMEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-50ASEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-150AEEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-150AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-250AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-150AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-250AEnvironmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Specifications
Cat. No.HI-5C8-100PEnvironmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-150PEnvironmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-250PEnvironmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-50PEnvironmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationLow Manufacturing Impact Specifications
Cat. No.HI-5C8-50SP1Environmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Specifications
Cat. No.HI-5C8-75PEnvironmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-100P3Environmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-150P3Environmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationLow Manufacturing Impact Specifications
Cat. No.HI-5C8-150SP1Environmentally PreferableLow Manufacturing ImpactCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Specifications
Cat. No.HI-5C8-250P3Environmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certificationSpecifications
Cat. No.HI-5C8-50P3Environmentally PreferableCarbon load8%EndcappedYesSurface area250 m²/gEnvironmental certification
Guard cartridges
- 説明カラム形式Phase細孔サイズ粒子サイズ内径長さ
Low Manufacturing Impact Specifications
Cat. No.HI-3.5C8-10C5Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.HI-5C8-10C5Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Specifications
Cat. No.HICHHI-5C8-10CPEnvironmentally PreferableCarbon loadEndcappedSurface areaEnvironmental certificationLow Manufacturing Impact Specifications
Cat. No.HI-5C8-10CP3Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedSurface areaEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard holders and accessories
- 説明
- PEEK coupler (for use with HI-161 or HI-150)1 * 1 items$0.00