Orders LinkContactUsLinkComponent
VWR®, オーバーオール、PP製、フードなし
VWR®, オーバーオール、PP製、フードなし
VWR®, オーバーオール、PP製、フードなし
Supplier:  VWR Collection
Certificates
VWR®, オーバーオール、PP製、フードなし
VWR®, オーバーオール、PP製、フードなし
4 Options Available Below
Product Details & Documents
Overalls made from non woven PP. Ideal for non critical applications.
  • Tapered collar, long sleeves and legs
  • Bound seams and zip fastening
  • Elasticated waist, wrist and ankles
Density: 40 g/m²
Colour: White
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8258
  • 113-8259
  • 113-8260
  • 113-8292
  • Model:
  • Zip
  • Zip
  • Zip
  • Zip
  • Garment application:
  • General Purpose|Industrial
  • General Purpose|Industrial
  • General Purpose|Industrial
  • General Purpose|Industrial
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    衣服のサイズ
    Availability
    Price
  • White
    L
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8258
    Model
    Zip
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8259
    Model
    Zip
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8260
    Model
    Zip
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²
  • White
    3XL
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8292
    Model
    Zip
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²