Orders LinkContactUsLinkComponent
VWR®, 作業着、PP、ディスポーザブル
VWR®, 作業着、PP、ディスポーザブル
VWR®, 作業着、PP、ディスポーザブル
VWR®, 作業着、PP、ディスポーザブル
Frocks made from PP, with tapered collar, and either press stud fastening or zip closure.
  • No pockets
  • Bound seams and elasticated wrists
Length for size XL: 1100 mm; 570 mm (sleeves).
Colour: White or blue
Weight: 40 g/m²
  • Catalog No:
  • 113-8269
  • 113-8295
  • 113-8296
  • 113-8270
  • 113-8294
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Zip closure
  • Zip closure
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Garment application:
  • Laboratory|Food
  • Laboratory|Food
  • Laboratory|Food
  • Laboratory|Food
  • Laboratory|Food
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Product Family Options
With press stud fastening
  • Colour
    衣服のサイズ
    長さ
  • White
    XL
    110 cm
    1 * 30 items
    1 * 30 items
    Specifications
    Cat. No.
    113-8269
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    Laboratory|Food
    Weight
    40 g/m²
  • Blue
    L
    110 cm
    1 * 30 items
    1 * 30 items
    Specifications
    Cat. No.
    113-8295
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    Laboratory|Food
    Weight
    40 g/m²
  • Blue
    XL
    110 cm
    1 * 30 items
    1 * 30 items
    Specifications
    Cat. No.
    113-8296
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    Laboratory|Food
    Weight
    40 g/m²
With zip closure
  • Colour
    衣服のサイズ
    長さ
  • White
    XL
    110 cm
    1 * 30 items
    1 * 30 items
    Specifications
    Cat. No.
    113-8270
    Closure type
    Zip closure
    Pockets
    Without pockets
    Garment application
    Laboratory|Food
    Weight
    40 g/m²
  • White
    XXL
    110 cm
    1 * 30 items
    1 * 30 items
    Specifications
    Cat. No.
    113-8294
    Closure type
    Zip closure
    Pockets
    Without pockets
    Garment application
    Laboratory|Food
    Weight
    40 g/m²