Orders LinkContactUsLinkComponent
VWR®, 拡散器、T字型
VWR®, 拡散器、T字型
VWR®, 拡散器、T字型
Supplier:  VWR Collection
Certificates
VWR®, 拡散器、T字型
VWR®, 拡散器、T字型
3 Options Available Below
Product Details & Documents
PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example, CFU) plate counts on water, milk and other liquid samples.
  • Smooth, rounded surfaces to prevent cutting or gouging of agar
  • T-shaped to ensure even pressure is applied while spreading
Product Family Options
Product Information
  • 説明
    Colour
    Packed
    Availability
    Price
  • Spreaders, T-shaped
    Blue
    1/peel pouch
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Spreaders, T-shaped
    Blue
    5/peel pouch
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Spreaders, T-shaped
    Blue
    10/peel pouch