Orders LinkContactUsLinkComponent
VWR®, スプレッダー、L字型、ディスポーザブル
VWR®, スプレッダー、L字型、ディスポーザブル
VWR®, スプレッダー、L字型、ディスポーザブル
Supplier:  VWR Collection
Certificates
VWR®, スプレッダー、L字型、ディスポーザブル
VWR®, スプレッダー、L字型、ディスポーザブル
2 Options Available Below
Product Details & Documents
PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Surface treated to enhance adhesion of droplets
  • Non toxic
  • Gamma irradiated
Dimensions: total length 145 mm (±1 mm); spreader length 38 mm (±0,5 mm).
Product Family Options
Product Information
  • 説明
    Colour
    Packed
    Availability
    Price
  • Spreaders, L-shaped
    Blue
    Individually packed
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Spreaders, L-shaped
    Blue
    5/bag