Orders LinkContactUsLinkComponent
VWR®, デジタルキャリパー
VWR®, デジタルキャリパー
Supplier:   VWR Collection
VWR®, デジタルキャリパー
Supplier:  VWR Collection
Certificates
VWR®, デジタルキャリパー
VWR®, デジタルキャリパー
2 Options Available Below
Product Details & Documents
Made from stainless steel. Battery life is about 10000 hours.
  • Ergonomic body and friction thumb roller make readings fast
  • Easy to read, jumbo digit display
  • Moisture and dust resistant steel
Zero button instantly sets closed jaws or any reference point to zero. Floating zero may also be set mid-measurement to read difference results without calculations.
Weight: 255 g.
納品情報: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by NIST.
Product Family Options
Product Information
  • 説明
    Range
    解像度
    Accuracy
    Availability
    Price
  • Digital caliper
    0 - 150 mm
    0,01 mm
    ±0,03 mm/mm
    1 * 1 items
  • Digital caliper
    0 - 200 mm
    0,01 mm
    ±0,03 mm/mm