VWR®, デスクトップ計算機
Desktop calculator HS-12 ER, 12 digit, 114×5×165 mm, 113,4 g
Catalog # 818-0066
Supplier: VWR Collection
Jump to:
Product Details & Documents
Tough ABS plastic case.
- Oversized keys
- Large digits
- 8- or 12-digit angled display
Arithmetic functions (add, subtract, multiply, divide), percent, three key memory, square root and floating decimal point.
With arithmetic functions (add, subtract, multiply, divide), percent, three key memory, square root, and floating decimal point.
Documents
Specifications
- Weight:113,4 g
- 説明:Desktop calculator HS-12 ER, 12 digit
- Seq:02
- W×D×H:114×5×165 mm