Orders LinkContactUsLinkComponent
VWR®, 割れたガラス用の箱
VWR®, 割れたガラス用の箱
VWR®, 割れたガラス用の箱
Supplier:  VWR Collection
Certificates
VWR®, 割れたガラス用の箱
VWR®, 割れたガラス用の箱
2 Options Available Below
Product Details & Documents
Corrugated cardboard with PE pouch, 50 µm thick.
  • Folded flat for space-saving storage
  • A flap is built into the lid for easy filling
  • The internal safety lid is closed for removal of the container
Ideal for collecting, transporting and removing broken glass from the laboratory. Consists of a sturdy box made of strong corrugated cardboard, and a PE insertion pouch which prevents any possible escape of fluids adhering to broken glass.
Does not qualify as a DOT container.
Delivery information: Delivered unassembled. Assembly takes a few seconds.
Product Family Options
Product Information
  • 説明
    W×D×H
    Availability
    Price
  • Floor standing box, max. weight: 18 kg
    305×305×685 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
  • Bench standing box, max. weight: 5,5 kg
    203×203×254 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
Accessories
Product Information
  • Catalog #
    説明
    For
    Colour
    Availability
    Price
  • VWRU89088-054
    Carton cover with bag holder
    Glass disposal carton
    Blue
    Log in for availability
    1 * 1 items - $63.35