Orders LinkContactUsLinkComponent
VWR®, ショウジョウバエ用ストックボトル
VWR®, ショウジョウバエ用ストックボトル
Supplier:   VWR Collection
VWR®, ショウジョウバエ用ストックボトル
Supplier:  VWR Collection
Certificates
VWR®, ショウジョウバエ用ストックボトル
VWR®, ショウジョウバエ用ストックボトル
4 Options Available Below
Product Details & Documents
PP, round or square bottom or PE, square bottom bottles.
  • Thin wall design for excellent visibility
  • Pliable top for easy removal of paper lid
  • Moulded-in food level graduation marks
Bottle 734-1259 features a wider edge for a more secure paper cap fit.
Bottle 734-1250 has a moulded indent near the bottom, which helps to secure media.
PP bottles are autoclavable, PE bottles are not.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • 説明
    容量
    Availability
    Price
  • PP, round bottom
    170 ml
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • PP, square bottom
    170 ml
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • PE, square bottom
    170 ml
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • PP, round bottom with moulded indent
    225 ml
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items