Digital Solutions
Applications
Support
About
Japan
Orders Link
ContactUsLinkComponent
クロージャー
Bottle Closures
交換用キャップ、VWR®製
交換用キャップ、VWR®製
:
VWR Chemicals
交換用キャップ、VWR®製
:
VWR Chemicals
交換用キャップ、VWR®製
Product Family Options
説明
29548.043
Cap with tap for 25 l (DIN 61) plastic carboys
1 * 1 items
-
+
1 * 1 items
-
+
223-0059
Cap with tap for 10 l plastic container
1 * 1 items
-
+
1 * 1 items
-
+
29548.075
Cap with pouring flex for 10 l (DIN 51) plastic carboys
1 * 1 items
-
+
1 * 1 items
-
+
29548.086
Cap with pouring flex for 20 or 25 l (DIN 61) plastic carboys